BriskHeat® Hazardous-Area T4-Rated Silicone Heating Blankets with Adhesive-120 VAC
FM approved for Class 1 Division 2 Groups A, B, C, and D, Class II Division 2, Groups F and G
  • Built-in high-limit thermostat keeps heater operating below designated NEC article T-rating
  • easy-to-apply industrial strength pressure sensitive adhesive backing
These industrial-strength heating blankets are designed for use in hazardous environments. They come equipped with a high-limit safety thermostat and 1-foot long power leads extending out of a 3/4” intrinsically-safe conduit fitting. The unique grounded heating element is laminated between two layers of extra strong 23 mil and two layers of 25 mil fiberglass reinforced silicone rubber. These heaters have a power density of 2.5 watts/in2 and should only be used on metal surfaces.
Approvals and compliances: FM, RoHS, REACH
Warning: Heating blankets should not be overlapped and requires properly rated temperature control and conduit.
84,300.00 - ₹138,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRX06243601PT4 Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 6x24" , 120V, 360W WW-36003-40 Mfr # SRX06243601PT4A
COMPARE
24 158 120
₹84,300.00INR / Each
BriskHeat SRX12123601PT4 Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 12x12" , 120V, 360W WW-36003-42 Mfr # SRX12123601PT4A
COMPARE
12 158 120
₹84,300.00INR / Each
BriskHeat SRX12247201PT4 Hazardous T4A-Rated Silicone Heating Blanket, Adhesive, 12x24" , 120V, 720W WW-36003-44 Mfr # SRX12247201PT4A
COMPARE
24 158 120
₹138,000.00INR / Each