This product is now discontinued, View Related Products

Haws Axion MSR Eyewash Bowls

Inverted flow sweeps contaminants away from inner eye and nasal cavity
Eyewashes feature a unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. With other eyewashes contaminants can travel through the inner corner of the eye to the nasal cavity resulting in further injury.

Bowls are 11" (27.9 cm) in diameter and include a push handle to start flow. Supply connection is 1/2" IPS; waste connection is 1-1/4" IPS.

Compliance: ANSI Z358.1-2009

Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Bowl Material Mount
Availability Pricing
Haws 7260B-7270B Eyewash Bowl, Wall-mount, ABS Plastic bowl WW-06767-31 Mfr # 7260B-7270B
COMPARE
ABS plastic Wall Mount
Call For Price
Haws 7360B-7460B Eyewash Bowl, Wall-mount, Stainless Steel bowl WW-06767-32 Mfr # 7360B-7460B
COMPARE
Stainless steel Wall Mount
Call For Price
Haws Axion MSR Eyewash Bowl, Wall-mount, ABS Plastic bowl with trap WW-06767-33 Mfr # 7260BT-7270BT
COMPARE
ABS plastic Wall Mount
Call For Price
Haws Axion MSR Eyewash Bowl, Wall-mount, Stainless steel bowl with trap WW-06767-34 Mfr # 7360BT-7460BT
COMPARE
Stainless steel Wall Mount
Call For Price
Haws 7261-7271 Eyewash Bowl, Pedestal, ABS Plastic bowl WW-06767-35 Mfr # 7261-7271
COMPARE
ABS plastic Pedestal Mount
Call For Price
Haws 7361-7461 Eyewash Bowl, Pedestal, Stainless steel bowl WW-06767-36 Mfr # 7361-7461
COMPARE
Stainless steel Pedestal Mount
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat