This product is now discontinued, View Related Products
Haws Axion MSR Combination Showers
Hydrodynamic shower design provides a more equal water distribution throughout the flow
  • Inverted flow sweeps contaminants away from inner eye and nasal cavity
Combination showers feature Axion MSR 11" (27.9 cm) diameter eyewash with unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. ABS plastic showerhead with flow control uses a hydrodynamic design to give a more equal distribution of water — provides the injured with a more thorough rinsing of the contaminant. Mesh water strainer reduces debris in the water supply for further eye protection. Constructed of galvanized steel pipe and fittings with a 9" (22.9 cm) cast iron floor flange.
Compliance: ANSI Z358.1-2009
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bowl Material
Availability Pricing
Haws 8320-8325 Combination Shower with ABS plastic Eyewash WW-06767-70 Mfr # 8320-8325
COMPARE
ABS plastic
Call For Price
Haws 8300-8309 Combination Shower with Stainless Steel Eyewash WW-06767-71 Mfr # 8300-8309
COMPARE
Stainless steel