Harvest Right Scientific Freeze Dryers
Customizable programming—ideal for drying in phases at different temperatures
  • User-friendly interface provides program customization— up to 10 thermal treatment and 12 drying steps
  • Constructed with a durable stainless steel chamber and collector
  • Large 4.3” full-color, touch-screen display provides real-time pressure and temperature data
  • Easily connect to a PC via ethernet cable for data logging and communication
These Harvest Right freeze dryers can be fully customized with the programming required for your application. This includes multiple manageable profiles, each providing the ability to automatically power on and off the condensing unit and the oil-free vacuum pump in conjunction with ascending, controlled temperature and time settings. Control the rate of freeze, ultimate final freeze temperature, and the duration of time your sample is held at the ultimate freeze temperature before the drying cycle begins. Control of the shelf temperature during the dry cycle, allowing the material to dry in phases, at different temperatures. Automatically control the rate of sublimation during the primary drying cycle with shelf heaters. Through this process, the unit has control of the attendant vacuum pressure. The freeze dryer records and logs the pressure and temperature during the entire freeze-dry cycle for ensured system performance. Connect the unit to a PC via Ethernet cable for data logging and exporting.
System includes a 7 CFM oil-free scroll vacuum pump which is quiet, fast, and does not require regular user maintenance.
1,809,000.00 - ₹2,412,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC) Description
Availability Pricing
Harvest Right Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Medium; 115 VAC 60 Hz WW-03336-51 Mfr # HRFD-Med-Sci
COMPARE
4 115 Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Medium; 115 VAC 60 Hz
₹1,809,000.00INR / Each
Harvest Right Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Medium; 220 VAC 50 Hz WW-03336-52 Mfr # HRFD-Med-Sci-EU
COMPARE
4 220 Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Medium; 220 VAC 50 Hz
₹1,809,000.00INR / Each
Harvest Right Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Large; 115 VAC 60 Hz WW-03336-53 Mfr # HRFD-Lrg-Sci
COMPARE
6 115 Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Large; 115 VAC 60 Hz
₹2,412,000.00INR / Each
Harvest Right Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Large; 220 VAC 50 Hz WW-03336-54 Mfr # HRFD-Lrg-Sci-EU
COMPARE
6 220 Scientific Freeze Dryer with Oil-Free Pump, Stainless Steel, Large; 220 VAC 50 Hz
₹2,412,000.00INR / Each