Tip Biosystems Handheld Photometer
Be mobile with this ultrafast portable spectrophotometer
  • Instant on-spot quantification of nucleic acids, proteins, cells density and many more
  • Robust optical system with precision ±0.02 AU
  • Geo tagging, photo, notes, date and time embedded in data file and real-time data sharing
  • Wireless connectivity to iOS/Android™ tablets and smartphones via app
  • Preprogrammed applications selectable by user
  • Unique disposable CuveTip® eliminates the need for sample transfer into a cuvette
  • No maintenance or lamp change required
The Photopette optical system is a multicomponent that integrates a photodiode detector with light emitting diodes (LEDs). It also has a built-in “environmental light correction” and can be used in the lab environment and outdoor without shielding environmental light from the sample. The photometer works together with a smartphone or computing device using a dedicated application which connects via a Bluetooth® link for taking measurements and data transfer.
The dual-purpose disposable reflective CuveTip was designed to act as a sample holder and a light-reflecting system. A minimum volume of 40 μL is required to fill the cavity of the CuveTip; this volume is recovered after the measurement.
Choose from four Photopette models. The Bio model allows direct measurement of DNA/RNA, protein concentration, and cell density. The Cell model permits fast measurement in the cell culture hood. The Turbidity model is ideal for wastewater and environmental monitoring applications. The Custom model lets you specify you three wavelengths.
Applications:
• Absolute quantification of cell density through calibration curve
• Mammalian, bacterial or yeast cell count
• Cell viability test using Resazurin dye
• Enzymatic assays for metabolites
• NAD/NADH based enzymatic assays
• DNA, RNA and Oligo concentration determination
• Bradford assay
• Direct protein concentration
• Lactate, pyruvate, glucose and alcohol
Designed for:
• Academic teaching laboratories (life science)
• Cell culture or tissue culture
• DNA and protein quantification
• Outdoor water and chemical analysis
• Farming and livestock
• Food and beverages
244,000.00 - ₹505,000.00INR / Case of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Wavelength (nm) Min Wavelength (nm) Description
Availability Pricing
Tip Biosystems PC-502 Handheld Photometer Cell System Kit, 340, 570, and 600 nm WW-16017-01 Mfr # 500008-PC
COMPARE
600 340 Handheld Photometer Cell System Kit, 340, 570, and 600 nm
₹256,000.00INR / Case of 1
Tip Biosystems PB-501 Handheld Photometer Bio System Kit, 260, 280, and 340 nm WW-16017-02 Mfr # 500008-PB
COMPARE
340 260 Handheld Photometer Bio System Kit, 260, 280, and 340 nm
₹281,000.00INR / Case of 1
Tip Biosystems PT-504 Handheld Photometer Turbidity System Kit, 570 and 860 nm WW-16017-03 Mfr # 500008-PT
COMPARE
860 570 Handheld Photometer Turbidity System Kit, 570 and 860 nm
₹278,000.00INR / Case of 1
Tip Biosystems PC-502 Handheld Photometer, Cell, Device Only WW-16017-04 Mfr # 500009-PC
COMPARE
600 340 Handheld Photometer, Cell, Device Only
₹388,000.00INR / Case of 1
Tip Biosystems PB-501 Handheld Photometer, Bio, Device Only WW-16017-05 Mfr # 500009-PB
COMPARE
600 260 Handheld Photometer, Bio, Device Only
₹471,000.00INR / Case of 1
Tip Biosystems PV-503 Custom Device with Custom Wavelengths WW-16017-07 Mfr # 500009-PV
COMPARE
1050 250 Custom Device with Custom Wavelengths
₹505,000.00INR / Case of 1
Tip Biosystems PT-504 Turbidity Kit WW-16017-10 Mfr # 500009-PT
COMPARE
860 570 Turbidity Kit
₹244,000.00INR / Case of 1
Showing 1- 7 of 7