Hamilton® Microliter Low-Volume Syringes
Achieve precise liquid delivery
  • Constructed of borosilicate glass
Hamilton® 700-series microliter syringes are designed to solve the general liquid handling requirements of manufacturing and research laboratories. Syringes are used for manual and automated GC and HPLC injections and for everyday applications that require accurate liquid measuring and dispending.
Choose from cemented needle, removable needle, or Luer tip syringes. Syringes with luer tip connections do not include a needle, order needles separately.
5,300.00 - ₹49,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Syringe Capacity (µL) Syringe Type
Availability Pricing
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 10 µL, 26s G WW-07938-00 Mfr # 80300
COMPARE
26s 10 Cemented Needle
₹5,300.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 25 µL, 22s G WW-07938-01 Mfr # 80400
COMPARE
22s 25 Cemented Needle
₹7,900.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 50 µL, 22s G WW-07938-02 Mfr # 80500
COMPARE
22s 50 Cemented Needle
₹9,700.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 100 µL, 22s G WW-07938-03 Mfr # 80600
COMPARE
22s 100 Cemented Needle
₹11,300.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 250 µL, 22s G WW-07938-04 Mfr # 80700
COMPARE
22s 250 Cemented Needle
₹13,400.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 500 µL, 22 G WW-07938-05 Mfr # 80800
COMPARE
22 500 Cemented Needle
₹14,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Small Hub Removable Need, 10 µL, 26s G WW-07938-10 Mfr # 80330
COMPARE
26s 10 Small hub removable needle
₹9,000.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Small Hub Removable Need, 25 µL, 22s G WW-07938-11 Mfr # 80430
COMPARE
22s 25 Small hub removable needle
₹11,500.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Small Hub Removable Need, 50 µL, 22s G WW-07938-12 Mfr # 80530
COMPARE
22s 50 Small hub removable needle
₹13,400.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Small Hub Removable Need, 100 µL, 22s G WW-07938-13 Mfr # 80630
COMPARE
22s 100 Small hub removable needle
₹14,900.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Large Hub Removable Need, 250 µL, 22s G WW-07938-14 Mfr # 80730
COMPARE
22s 250 Large hub removable needle
₹17,000.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Large Hub Removable Need, 500 µL, 22 G WW-07938-15 Mfr # 80830
COMPARE
22 500 Large hub removable needle
₹18,200.00INR / Each
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 25 µL WW-07938-21 Mfr # 80401
COMPARE
Needle not included 25 Luer Tip
₹12,000.00INR / Each
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 50 µL WW-07938-22 Mfr # 80501
COMPARE
Needle not included 50 Luer Tip
₹8,900.00INR / Each
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 100 µL WW-07938-23 Mfr # 80601
COMPARE
Needle not included 100 Luer Tip
₹10,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 250 µL WW-07938-24 Mfr # 80701
COMPARE
Needle not included 250 Luer Tip
₹12,500.00INR / Each
Hamilton Microliter Low-Volume Syringe, Luer Tip, No Needle, 500 µL WW-07938-25 Mfr # 80801
COMPARE
Needle not included 500 Luer Tip
₹14,200.00INR / Each
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Cemented Needle, 10 µL, 26s G; 6/Pk WW-07938-30 Mfr # 80366
COMPARE
26s 10 Cemented Needle
₹29,300.00INR / Pkg of 6
Hamilton Microliter Low-Volume Syringe, Beveled Tip, Small Hub Removable Need, 10 µL, 26s G; 6/Pk WW-07938-40 Mfr # 80336
COMPARE
26s 10 Small hub removable needle
₹49,000.00INR / Pkg of 6
Showing 1- 19 of 19