Hamilton® 7000-Series Microliter Low-Volume Syringes with Needle
Designed with zero dead volume
  • Gas- and liquid-tight up to 6000 psi!
Hamilton® 7000-series microliter syringes are designed for rapid injection of minute volumes. Full syringe capacity is in the needle and is completely ejected when plunger wire is depressed—no void volume. The fine gauge plunger wire extends all the way to the tip of the needle ensuring that all solvent is expelled from the needle. Therefore, it is not recommended to remove the needle or plunger.
18,600.00 - ₹18,700.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Needle Tip Style Syringe Capacity (µL)
Availability Pricing
Hamilton Microliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 0.5 µL, 25 G WW-07938-60 Mfr # 86250
COMPARE
25 90⁰ Blunt Tip 0.5
₹18,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 1 µL, 25 G WW-07938-61 Mfr # 80100
COMPARE
25 90⁰ Blunt Tip 1
₹18,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 2 µL, 25 G WW-07938-62 Mfr # 88400
COMPARE
25 90⁰ Blunt Tip 2
₹18,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Knurled Hub, Blunt Tip, 5 µL, 24 G WW-07938-63 Mfr # 88000
COMPARE
24 90⁰ Blunt Tip 5
₹18,600.00INR / Each
Hamilton Microliter Low-Volume Syringe, Knurled Hub, Beveled Tip, 1 µL, 25 G WW-07948-61 Mfr # 80135
COMPARE
25 Beveled Tip 1
₹18,700.00INR / Each