This product is now discontinued, View Related Products
H-B Instrument Enviro-Safe Nonhazardous Dry Block Glass Thermometers, Celsius-Scale
Replace mercury thermometers for Dry Block applications with the same accuracy
  • 100% non-toxic, non-hazardous, biodegradable, and “green”
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy
Designed for use in all standard heating blocks and water baths. All Enviro-Safe glass thermometers contain an environmentally friendly nonmercury liquid that is 100% certified as nontoxic, nonhazardous, and fully biodegradable. In case of breakage, the liquid can be cleaned up easily with soap and water. These thermometers feature a green liquid fill, white back glass, arrows indicating critical temperatures, and individual serial numbers. Come with a statement of accuracy and recyclable packaging.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60510-0000 Enviro-Safe Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill WW-08006-07 Mfr # 605100000
COMPARE
57 24 175
Call For Price
H-B Instrument B60510-0100 Enviro-Safe Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, PFA Coated, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill WW-08006-08 Mfr # 605100100
COMPARE
57 24 175