H-B Instrument Easy-Read Enviro-Safe Nonhazardous Glass Tank Thermometers Traceable to NIST
Supplied with ASTM "Like" approved, non-mercury thermometers
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Supplied with ASTM "Like" approved, non-mercury thermometers
  • Thermometers can be remounted can be replaced if breakage occurs
All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Description
Availability Pricing
H-B Instrument B60380-0300 Easy-Read Liquid-In-Glass Tank Thermometer, -20 to 110°C, 76 mm Immersion, Environmentally Friendly WW-93902-03 Mfr # 603800300
COMPARE
110 230 Easy-Read Liquid-In-Glass Tank Thermometer; -20 to 110C, 76mm Immersion, Environmentally Friendly
Call For Price
H-B Instrument B60380-0500 Easy-Read Liquid-In-Glass Tank Thermometer, 0 to 230°F, 76 mm Immersion, Environmentally Friendly WW-93902-05 Mfr # 603800500
COMPARE
110 230 Easy-Read Liquid-In-Glass Tank Thermometer; 0 to 230F, 76mm Immersion, Environmentally Friendly