Cole-Parmer Durac Polycarbonate Salt Hydrometers, plain form
These durable polycarbonate hydrometers won''t break when dropped—perfect for industrial or laboratory use.
The temperature standard for these hydrometers is 60°F (15°C). All hydrometers are individually serialized, feature metal ballast with wax epoxy and include multi-language statement of accuracy supplied by the vendor.
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
22,000.00 - ₹25,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Salt Brine Shatterproof Plastic Hydrometer; 0/80 Percent by Saturation
₹25,000.00INR / Each
COMPARE
Salt Brine Shatterproof Plastic Hydrometer; 10/100 Percent by Saturation
₹22,000.00INR / Each