Cole-Parmer Calibrated API ASTM Plain Form Hydrometers
No more calibration delays
  • Plain form simplifies readings
  • Individually serialized and calibrated at three points for superior accuracy out of the box
  • Lead-free ballast and glass construction is nontoxic
  • Temperature standard is 60°F (15.6°C)
133,000.00 - ₹134,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Individually Calibrated 19/31 Degree API ASTM 3H Hydrometer
₹133,000.00INR / Each
COMPARE
Individually Calibrated 29/41 Degree API ASTM 4H Hydrometer
₹134,000.00INR / Each