Cole-Parmer Calibrated Alcohol Proof Glass Hydrometers
Determine percent of alcohol of waste liquors
  • Temperature standard is 60°F (15.6°C)
  • Ensure accuracy with multi-language statement of accuracy supplied by manufacturer
Available in various ranges from 30 to 170%, the calibrated alcohol proof glass hydrometers are ideal for measuring the percent of alcohol of waste liquors. The hydrometers feature lead-free ballast and glass.
23,900.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
150/170 Percent Alcohol Proof Hydrometer
₹23,900.00INR / Each