This product is now discontinued, View Related Products
GW Instek GDM 8300 Series Dual-Display/Dual-Measurement Multimeters
Conveniently save and retrieve data
  • User-selectable measurement speed maximizes measurement efficiency and accuracy
  • Dual display provides multiple measurement visibility
  • 0.02% DC voltage accuracy provides precision measurements
  • USB device standard, with optional USB host and GPIB for additional flexibility
The GDM-8300 series is ideal for manufacturing process tests, testing facilities, educational experiments, and more. Units feature USB flash drive storage functionality allowing you to download data without connecting to a PC. Model 8342 also provides a USB interface (virtual COM port) for downloading data when connected to a PC, and model 8342GP offers an additional GPIB interface for automating the unit and conveniently saving and retrieving data.
Additional functions include two-wired resistance, maximum/minimum values, reading hold, relative values, dB, dBm, algorithms (MX+B, 1/X, Ref%), comparison. Model 8342 also offers temperature measurement using thermocouple input (type J, K, or T).
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek GDM-8341 Multimeter, Benchtop, 50 k Counts, with USB Device WW-20050-43 Mfr # GDM-8341
COMPARE
Multimeter, Benchtop, 50 k Counts, with USB Device
Call For Price
GW Instek GDM-8342 Multimeter, Benchtop, 50 k Counts, with USB Host/Device WW-20050-44 Mfr # GDM-8342
COMPARE
Multimeter, Benchtop, 50 k Counts, with USB Host/Device
Call For Price
GW Instek GDM-8342GP Multimeter, Benchtop, 50 k Counts, with USB Host/Device and GPIB WW-20050-45 Mfr # GDM-8342GP
COMPARE
Multimeter, Benchtop, 50 k Counts, with USB Host/Device and GPIB