This product is now discontinued, View Related Products
GW Instek GDM-8261A 6-1/2 Digit Dual-Measurement Benchtop Multimeter
Increase measurement speed and efficiency
  • Perform two selected measurements simultaneously
  • Models with GPIB or LAN card option offer additional flexibility
  • Optional scanner card eliminates the complexities of software development for multipoint measurement and data processing
  • Two- and four-wire measurement methods provide the most accurate measurement results
  • RTD and thermocouple measurement capability for a wide range of temperature applications
  • Free PC software and driver lets you remotely control unit without having to develop programming software
The GDM-8261A high-precision multimeter features multipoint measurement capability, allowing you to use the unit as a semi-automated ATE system to increase the throughput of manufacturing test, or to easily configure as a data logger to perform long-term monitoring or characterization of a device under test. Store and recall up to 9999 measurement readings.
Unit features eleven measurement functions and ten math functions with precision accuracy to accommodate the most frequently performed parameter measurements, covering a wide range of application requirements.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek GDM-8261A Multimeter, Benchtop, 6.5" Digit WW-20050-50 Mfr # GDM-8261A
COMPARE
Multimeter, Benchtop, 6.5\" Digit
Call For Price
GW Instek GDM-8261A-OPT2 Multimeter, Benchtop, 6.5" Digit, with GPIB Card WW-20050-51 Mfr # GDM-8261A-OPT2
COMPARE
Multimeter, Benchtop, 6.5\" Digit, with GPIB Card
Call For Price
GW Instek GDM-8261A-OPT3 Multimeter, Benchtop, 6.5" Digit, with LAN Card WW-20050-52 Mfr # GDM-8261A-OPT3
COMPARE
Multimeter, Benchtop, 6.5\" Digit, with LAN Card