GW Instek DC Milliohm Meters
Ideal for low-resistance measurements
  • Obtain measurements quickly with a 60 reading per second sampling rate and 50,000 counts display
  • Easily read results on the 3.5" (320 x 240) TFT LCD
  • Depend on high accuracy and resolution — with ±0.05% precision and 1µΩ resolution
  • Connect to an external sorting machine, external on/off switch, or trigger control using the communication ports
These DC milliohm meters come loaded with great features ideal for your low resistance measurement needs. Depend on high accuracy and resolution even when using measurement speeds up to 60 samples per minute — the color LCD will always display five digits.
251,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek GOM-805 DC Milliohm Meter with GPIB and Dry Circuit; 100-240 V, 50/60 Hz WW-20025-95 Mfr # GOM-805
COMPARE
DC Milliohm Meter with GPIB and Dry Circuit; 100-240 V, 50/60 Hz
₹251,000.00INR / Each