This product is now discontinued, View Related Products
GW Instek AFG-3000 Series Arbitrary Function Generators
Simplify your diversified waveform needs
  • Frequency resolution throughout the full range provides precision accuracy
  • Direct digital synthesis technology generates precision waveforms
  • Ten sets of memory allow quick and easy saving and recalling of waveforms
  • Direct Waveform Reconstruction (DWR) function lets you easily edit waveforms and enhance measurement efficiency
  • User-selectable impedance function ensures the correct impedance compatibility
The AFG-3000 series of arbitrary waveform generators is designed for industrial, scientific research, and educational applications. Units feature 16-bit high resolution, 200 MS/s sampling rate, and 1 M memory, allowing you to easily edit arbitrary waveforms through the intuitive user interface.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek AFG-3051 Function Generator, 50 MHz WW-20050-53 Mfr # AFG-3051
COMPARE
Function Generator, 50 MHz
Call For Price
GW Instek AFG-3081 Function Generator, 80 MHz WW-20050-54 Mfr # AFG-3081
COMPARE
Function Generator, 80 MHz