Guardian Equipment Combination Safety Showers with Eyewash
Enhance safety with combination safety stations that feature a shower and eyewash bowl
  • Safety stations feature a 10” diameter shower head
  • GS-Plus™ spray heads provide flow control and filter to remove impurities from water
  • Easily visible ANSI-compliant identification sign
  • Furnished with orange polyethylene pipe covers for high visibility and corrosion resistance
These Guardian Equipment combination safety stations feature a free-standing shower and an eyewash station for optimal safety. All stations feature a stay open ball valve in both the shower head and wash bowl. Furnished with a stainless steel actuating arm and pull rod for shower operation.
Choose from models with a stainless steel or an ABS plastic shower head or bowl construction, hand only or hand and foot operation, and standard or WideArea™ for improved access.
205,000.00 - ₹508,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Eyewash Fixture Material Description
Availability Pricing
Guardian Equipment Combination Safety Shower with Eyewash, Barrier-Free Shower with WideArea™ Eye/Face Wash, Stainless Steel Bowl and ABS Plastic Shower Head WW-01850-58 Mfr # GBF1909
COMPARE
Stainless Steel Chrome-Plated Brass Combination Safety Shower with Eyewash, Barrier-Free Shower with WideArea™ Eye/Face Wash, Stainless Steel Bowl and ABS Plastic Shower Head
₹251,000.00INR / Each
Guardian Equipment Combination Safety Shower with Eyewash, Stainless Steel Bowl and Shower Head WW-01850-59 Mfr # G1991
COMPARE
Stainless Steel 316 Stainless Steel Combination Safety Shower with Eyewash, Stainless Steel Bowl and Shower Head
₹508,000.00INR / Each
Guardian Equipment Combination Safety Shower with Eyewash, ABS Plastic Bowl and Shower Head WW-06767-40 Mfr # G1902P
COMPARE
ABS plastic Chrome-Plated Brass Combination Safety Shower with Eyewash, ABS Plastic Bowl and Shower Head
₹205,000.00INR / Each
Guardian Equipment Combination Safety Shower with Eyewash, Hand and Foot Control, ABS Plastic Bowl and Shower Head WW-06767-41 Mfr # G1902P-HFC
COMPARE
ABS plastic Chrome-Plated Brass Combination Safety Shower with Eyewash, Hand and Foot Control, ABS Plastic Bowl and Shower Head
₹223,000.00INR / Each
Guardian Equipment Combination Safety Shower with Eyewash, Stainless Steel Bowl and ABS Plastic Shower Head WW-06767-43 Mfr # G1902
COMPARE
Stainless Steel Chrome-Plated Brass Combination Safety Shower with Eyewash, Stainless Steel Bowl and ABS Plastic Shower Head
₹228,000.00INR / Each
Guardian Equipment Combination Safety Shower with Eyewash, Stainless Steel Bowl and Cover and ABS Plastic Shower Head WW-06767-45 Mfr # G1902BC
COMPARE
Stainless Steel Chrome-Plated Brass Combination Safety Shower with Eyewash, Stainless Steel Bowl and Cover and ABS Plastic Shower Head
₹285,000.00INR / Each