Guardian AutoFlow Wall-Mount Eyewash
Simply pull down the sprayhead to activate flow
  • Inline strainer protects sprayheads from debris in the water line
Install this wall-mount eyewash above your sink or install above a floor drain to prevent water accumulation. Features two GS-Plus™ sprayheads with a flip-top dust cover, internal flow control, and filter. Eyewash actuates upon pulling the assembly down and remains in operation until it is pushed back into the storage position. Water supply inlet is 1/2" NPT(F).
Compliance: ANSI Z385.1-2009
137,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Eyewash Fixture Material
Availability Pricing
Guardian Equipment G1848LH-L AutoFlow Wall-Mount Eyewash, Chrome-plated brass WW-06767-11 Mfr # G1848LH-L
COMPARE
Chrome-plated brass
₹137,000.00INR / Each