GT Scien TOGA® Toxic Gas Purification Systems for Flammable Storage
A cutting-edge solution designed to purify harmful gases emitted from chemicals stored in flammable cabinets
  • Advanced filtration provides a state-of-the-art system for safe gas purification
  • Custom TOGA® filter is tailored for effective removal of harmful gases
  • Can be used as a tabletop fume extractor for added convenience
  • Prevents side reactions and harmful emissions for safety
  • Digital model adds on remote control, web service, and an intuitive UX display
Introducing the TOGA® toxic gas purifier module, a groundbreaking solution for laboratories and facilities dealing with chemicals stored in flammable cabinets. This toxic gas purifier employs an advanced filtration system, featuring custom TOGA® filters to effectively remove harmful gases. The digital model boasts remote control capabilities, utilizing SmartLab Mate AI for efficient operation. Real-time monitoring is made possible through the LSPMS Web Service, offering interactive graphs for temperature control of the digital model. Technical specifications include compact dimensions, a user-friendly 4.3-inch touch screen on the digital model or on/off switch on the analog model, and a powder coating for excellent corrosion resistance. With CPVC piping ensuring chemical resistance, this purifier enhances safety by preventing harmful emissions and extending the life of flammable cabinets.
331,000.00 - ₹551,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Type Fixtures Power (VAC)
Availability Pricing
GT SCIEN TOGA® Toxic Gas Purification System for Flammable Storage; 120V WW-81790-53 Mfr # TOGA-SCA0P
COMPARE
TOGA® On/Off switch 120
₹331,000.00INR / Each
GT SCIEN TOGA® Digital Toxic Gas Purification System for Flammable Storage; 120V WW-81790-54 Mfr # TOGA-SCD0P
COMPARE
TOGA® 4.3 in Touch screen 120
₹551,000.00INR / Each