BriskHeat® Grounded Heating Tapes - 115 VAC
Safe for heating metal vessels
  • Heat to 482F (250C)
  • NOTE: Temperature range has been reduced to meet safty standards
Wind heating tape around glass, ceramic, or metal vessels—grounding prevents shock hazard. Temperature-resistant fiberglass sleeves encase heating element; ground wire at core. The maximum surface loading is 4 watts/sq in. Leads are 31/2" long with electrical connector at one end. Tapes include ties at both ends.
WARNING: Heating tape should not be overlapped and require the use of a temperature or power controller sold separately.
15,400.00 - ₹39,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat CPBIH101020LGT Grounded Fiberglass Insulated Heat Tape, 2' x 1" , 100 Watts, 115 V WW-03121-10 Mfr # CPBIH101020LGT
COMPARE
2 482 115
₹15,400.00INR / Each
BriskHeat CPBIH101030LGT Grounded Fiberglass Insulated Heat Tape, 3' x 1" , 150 Watts, 115 V WW-03121-20 Mfr # CPBIH101030LGT
COMPARE
3 482 115
₹17,000.00INR / Each
BriskHeat CPBIH101040LGT Grounded Fiberglass Insulated Heat Tape, 4' x 1" , 200 Watts, 115 V WW-03121-30 Mfr # CPBIH101040LGT
COMPARE
4 482 115
₹18,700.00INR / Each
BriskHeat CPBIH101060LGT Grounded Fiberglass Insulated Heat Tape, 6' x 1" , 300 Watts, 115 V WW-03121-40 Mfr # CPBIH101060LGT
COMPARE
6 482 115
₹23,000.00INR / Each
BriskHeat CPBIH101080LGT Grounded Fiberglass Insulated Heat Tape, 8' x 1" , 400 Watts, 115 V WW-03121-50 Mfr # CPBIH101080LGT
COMPARE
8 482 115
₹26,300.00INR / Each
BriskHeat CPBIH101016LGT Grounded Fiberglass Insulated Heat Tape, 16' x 1" , 800 Watts, 115 V WW-03121-70 Mfr # CPBIH101016LGT
COMPARE
16 482 115
₹39,800.00INR / Each