Cole-Parmer® Graphite Short Ferrules Compatible with Agilent Inlets
Reusable one-piece design
  • Constructed of durable graphite
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450°C. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.
5,200.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16\" to 0.4 mm
₹5,200.00INR / Pkg of 10
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16\" to 0.5 mm
₹5,200.00INR / Pkg of 1
COMPARE
Graphite Short Ferrules Compatible with Agilent Inlets, 1/16\" to 0.8 mm
₹5,200.00INR / Pkg of 1