Corning® Gosselin™ Transpartent Autoclavable Biohazard Waste Bags
Safely decontaminate waste in your autoclave
  • Thick polypropylene (PP) bags handles lab waste
  • Bags are available with a biohazard logo
Gosselin autoclavable PP bags are 40 µm thick—strong enough to hold your lab waste during the autoclaving process. Bags withstand temperatures up to 273°F (134°C).
49,400.00INR / Case of 500
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Thickness (mil) Volume (Liters)
Availability Pricing
Corning Gosselin Autoclavable Bags, 9L, polypropylene, with biohazard symbol; 500/cs WW-34551-94 Mfr # SA09-10
COMPARE
PP 0.04 9
₹49,400.00INR / Case of 500