Corning® Gosselin™ Petri Dishes, Polystyrene
Optimal flatness ensures constant depth of agar
  • Vented dishes reduce condensation and allow air circulation
  • Class 100 manufacturing environment ensures aseptic production and packaging
These petri dishes are constructed of crystal polystyrene with an injection point located on the side of the dish bottom to optimize clarity even by image analyzing systems. Dishes are available in slippable or stackable designs, sterile or aseptic, and general lab or traceable clean packaging.
Slippable dishes are automation-compatible while stackable dishes have a stacking ring which makes them practical to pile.
Dishes are available in aseptic and sterile versions. Aseptic products are manufactured in a controlled environment, without human intervention during the manufacturing process. Sterile products are sterilized using an irradiation method compliant with the requirements of ANSI/AAMI/ISO 11137 to achieve (SAL 10⁻⁶).
General lab packaging does not have an outer bag, making it more economic and environmentally friendly. Clean packaging features two outer bags and bar code label for optimal traceability and suitable for clean room use.
19,200.00 - ₹31,000.00INR / Case of 500
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Closure Packaging Sterile
Availability Pricing
Corning Gosselin Petri Dish, Polystyrene, General Lab Packaging, Slippable, Sterile, 4 Vents, 100 x 15 mm; 500/Cs WW-34551-68 Mfr # BP94A-01
COMPARE
4 Vents; Slippable General Lab Packaging Sterile
₹19,200.00INR / Case of 500
Corning Gosselin Petri Dish, Polystyrene, General Lab Packaging, Stackable, Sterile, 4 Vents, 100 x 15 mm; 500/Cs WW-34551-69 Mfr # BP94S-01
COMPARE
4 Vents; Stackable General Lab Packaging Sterile
₹19,200.00INR / Case of 500
Corning Gosselin Petri Dish, Polystyrene, Clean Packaging, Stackable, Sterile, 3 Vents, 150 x 15 mm; 176/Cs WW-34551-80 Mfr # BP143-06
COMPARE
3 Vents; Stackable Clean Packaging Sterile
₹31,000.00INR / Case of 176
Corning Gosselin Petri Dish, Polystyrene, General Lab Packaging, Stackable, Aseptic, 3 Vents, 100 x 15 mm; 825/Cs WW-34551-82 Mfr # SB93-101
COMPARE
3 Vents; Stackable General Lab Packaging Aseptic
₹27,800.00INR / Case of 825
Corning Gosselin Petri Dish, Polystyrene, Clean Packaging, Stackable, Sterile, 3 Vents, 100 x 15 mm; 825/Cs WW-34551-83 Mfr # BP93B-102
COMPARE
3 Vents; Stackable Clean Packaging Sterile
₹29,800.00INR / Case of 825
Corning Gosselin Petri Dish, Polystyrene, Clean Packaging, Stackable, Aseptic, 3 Vents, 100 x 15 mm; 825/Cs WW-34551-86 Mfr # BP93B-101
COMPARE
3 Vents; Stackable Clean Packaging Aseptic
₹26,900.00INR / Case of 825