Glove Box Sleeves
Ideal for use with neoprene or latex glove box hands
These neoprene straight sleeves are ideal for use with neoprene or latex glove box hands. The sleeves fit 8 to 8-3/4" openings.
Order glove box hands separately.
113,000.00INR / Pair of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Transfer Chamber Length (in) Description
Availability Pricing
Labconco 5005800 Glove Box Sleeves, Straight, 28" Long with Hands, Neoprene; 1 Pair WW-09112-18 Mfr # 5005800
COMPARE
28 Glove Box Sleeves, Straight, 28\" Long with Hands, Neoprene; 1 Pair
₹113,000.00INR / Pair of 1