Analtech Glass Thin Layer Chromatography (TLC) Developing Tanks and Racks
Designed for the development of TLC plates
  • Tanks are constructed of durable glass and racks are constructed of anodized aluminum
Tanks accommodate standard TLC plates. The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development. Thinline™ tanks accommodate two plates and require less solvent for development. Latch-Lid™ units hold lids in place with stainless steel latches. Order rack to develop maximum number of plates in a tank.
23,700.00 - ₹72,300.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Plate Capacity Plate Length (cm) Plate Width (cm)
Availability Pricing
Analtech TLC Developing Tank with LatchLid™ and Aluminum Rack, Glass, Holds Six 10 x 10 cm Plates WW-34105-05 Mfr # A70-25
COMPARE
6 10 10
₹72,300.00INR / Each
Analtech TLC Developing Tank with LatchLid™, Glass, Holds Six 10 x 10 cm Plates WW-34105-10 Mfr # A70-29
COMPARE
6 10 10
₹39,300.00INR / Each
Analtech TLC Developing Tank with Lid, Glass, Holds Six 10 x 10 cm Plates WW-34105-15 Mfr # A70-30
COMPARE
6 10 10
₹23,700.00INR / Each
Analtech TLC Developing Tank with LatchLid™, Glass, Holds Five 20 x 20 cm Plates WW-34105-20 Mfr # A80-33
COMPARE
5 20 20
₹70,400.00INR / Each
Analtech Rack for TLC Developing Tank, Anodized Aluminum, Holds Six 20 x 20 cm Plates WW-34105-25 Mfr # A80-31
COMPARE
5 20 20
₹45,200.00INR / Each
Analtech Rack for TLC Developing Tank, Anodized Aluminum, Holds Six 10 x 40 cm Plates WW-34105-40 Mfr # A70-28
COMPARE
6 10 10
₹49,500.00INR / Each
Analtech TLC Developing Tank with Lid, Glass, Holds Five 20 x 20 cm Plates WW-34105-50 Mfr # A70-22
COMPARE
5 20 20
₹34,800.00INR / Each
Showing 1- 7 of 7