Glass Gas Sampling Bulb
Glass Gas Sampling Bulbs are designed for the collection and storage of environmental samples. Top cap with a PTFE® PTFE/silicone septum lets you withdraw analytical samples. Both ends have a 4-mm valve plug and are ground to accept 3⁄8" ferrules and hand compressible fittings for a gas-tight seal. GPI thread size: 13-425.
60,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters)
Availability Pricing
DWK Life Sciences (Kimble) 653150-0125 Gas Sampling Bulb, glass, 125 mL, 1/ea WW-10370-00 Mfr # 653150-0125
COMPARE
0.125
₹60,100.00INR / Each