GHS Exploding Bomb Labels
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exploding Bomb pictogram indicates the following potential hazard: explosives, self-reactives, and organic peroxides.
5,600.00INR / Roll of 250
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH603EV2 GHS Label,Exploding Bomb Pictogram,1" x 1",adhesive poly,250/roll WW-50712-66 Mfr # LZH603EV2
COMPARE
1 Adhesive Poly 250/roll
₹5,600.00INR / Roll of 250