General Tools MM1E Moisture Meter with LED Bar Graph
A must-have tool for woodworking, building construction, and home inspection
  • Check for moisture in carpets and subflooring
  • Great for identifying leaks in roofs, floors, and walls
  • 10-LED dual-scale readout with separate High/Low indicators
Use this low-cost meter with pins for a surface moisture measurement in building materials such as wood, plaster, drywall, concrete, roofing, and insulation. A protective cap doubles as a calibration checker. Pins provide 3/8” (1 cm) penetration for testing soft materials such as soil, paper, or powders. Simply replace the pins when they wear out. Unit features full- and low-battery indicators.
3,200.00 - ₹10,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Moisture Range Non-Wood Scale (Max) (%) Moisture Range Wood Scale (Max) (%) Description
Availability Pricing
General Tools MM1E Moisture Meter with LED Bar Graph WW-37800-13 Mfr # MM1E
COMPARE
35 Moisture Meter with LED Bar Graph
₹10,200.00INR / Each
General Tools PIN4E Replacement Pins for MM1E and MMH800 (2 Pair) WW-37800-19 Mfr # PIND4E
COMPARE
Replacement Pins for MM1E and MMH800 (2 Pair)
₹3,200.00INR / Each