This product is now discontinued, View Related Products

GasAlertMicro 5 Multigas Detectors

Monitors and displays up to five potential atmospheric hazards
  • Full function self-test of: sensor, battery and circuit integrity; and audible/ visual alarms
  • Compact and lightweight
  • Rugged, highly water-resistant design
  • Continuous LCD shows simultaneous gas concentrations for up to five gases
  • Integral concussion-proof enclosure for the most rugged environments
  • Integral motorized pump option for remote sampling (optional)
  • Two power options: three AAA alkaline batteries or a rechargeable battery pack
  • Provides 95dB alarm tone (typical), two bright wide-angled alarm bars and internal vibrating alarm for high-noise areas
  • Multi-language supporting English, French, German, Spanish and Portuguese
  • Four alarm levels: instantaneous Low and High alarm for all gases; TWA (time weighted average) and STEL (short term exposure limit) for toxic gases
  • Simple automatic calibration procedure: compatible with MicroDock II system
  • Records TWA, STEL and maximum exposures to gas, and displays readings on command
  • The GasAlertMicro 5 has user-selectable field options which include: confidence beep, pass code protection, latching alarm, “SAFE” display function, stealth mode, span level, and selectable ppm resolution for some gases
GasAlertMicro 5 portable gas detector simultaneously monitors and displays up to five potential atmospheric hazards including oxygen, combustible gases and a range of toxic gases. Compact, lightweight and water-resistant. Audible, visual and vibrator alarms activate in the event of a low, high, TWA or STEL alarm condition. An optional motorized pump is available.
Call For Price

24 variations of this product are available.

See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Battery Type Gases Measured Description
Availability Pricing
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, NH3, Combustibles, PID WW-10106-99 Mfr # M5PID-XWQA-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, NH3, Combustibles, PID
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/NH3/LEL/PID, Data Logger, Pump WW-10107-00 Mfr # M5PID-XWQA-R-P-D-Y-N-00
COMPARE
Rechargable battery Ammonia, combustibles, O2, PID Multigas Detector; O2/NH3/LEL/PID, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, NH3, Cl2, Combustibles WW-10107-02 Mfr # M5-XWCA-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, NH3, Cl2, Combustibles
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector: O2, NH3, Cl2, Combustibles; with data logger WW-10107-03 Mfr # M5-XWAC-R-P-D-Y-N-00
COMPARE
Rechargable battery Ammonia, chlorine, combusitbles, O2 Multigas Detector: O2, NH3, Cl2, Combustibles; with data logger
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, Cl2, Combustibles, PID WW-10107-05 Mfr # M5PID-XWQC-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, Cl2, Combustibles, PID
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/Cl2/LEL/PID, Data Logger, Pump WW-10107-22 Mfr # M5PID-XWQC-R-P-D-Y-N-00
COMPARE
Rechargable battery Chlorine, combustibles (LEL), O2, PID Multigas Detector; O2/Cl2/LEL/PID, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, Cl2, H2S, CO, Combustibles WW-10107-24 Mfr # M5-XWCY-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, Cl2, H2S, CO, Combustibles
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/Cl2/H2S/CO/LEL, Data Logger, Pump WW-10107-25 Mfr # M5-XWCY-R-P-D-Y-N-00
COMPARE
Rechargable battery CO, chlorine, combustibles (LEL), H2S, O2 Multigas Detector; O2/Cl2/H2S/CO/LEL, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, H2S, CO, Combustibles WW-10107-51 Mfr # M5-XW0Y-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, H2S, CO, Combustibles
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector: O2, H2S, CO, Combustibles; with data logger WW-10107-52 Mfr # M5-XW0Y-R-P-D-Y-N-00
COMPARE
Rechargable battery CO, combustibles, H2S, O2 Multigas Detector: O2, H2S, CO, Combustibles; with data logger
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, PH3, Combustibles, PID WW-10107-53 Mfr # M5PID-XWQP-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, PH3, Combustibles, PID
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/PH3/LEL/PID, Data Logger, Pump WW-10107-54 Mfr # M5PID-XWQP-R-P-D-Y-N-00
COMPARE
Rechargable battery Combustibles (LEL), O2, PID, phosphane Multigas Detector; O2/PH3/LEL/PID, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, PH3, CO, H2S, Combustibles WW-10107-57 Mfr # M5-XWPY-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, PH3, CO, H2S, Combustibles
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/PH3/CO/H2S/LEL, Data Logger, Pump WW-10107-58 Mfr # M5-XWPY-R-P-D-Y-N-00
COMPARE
Rechargable battery CO, combustibles (LEL), H2S, O2, phosphane Multigas Detector; O2/PH3/CO/H2S/LEL, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, SO2, Combustibles, PID WW-10107-60 Mfr # M5PID-XWQS-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, SO2, Combustibles, PID
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/SO2/LEL/PID, Data Logger, Pump WW-10107-61 Mfr # M5PID-XWQS-R-P-D-Y-N-00
COMPARE
Rechargable battery Combustibles (LEL), O2, PID, SO2 Multigas Detector; O2/SO2/LEL/PID, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, SO2, CO, H2S, Combustibles WW-10107-64 Mfr # M5-XWSY-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, SO2, CO, H2S, Combustibles
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/SO2/CO/H2S/LEL, Data Logger, Pump WW-10107-65 Mfr # M5-XWSY-R-P-D-Y-N-00
COMPARE
Rechargable battery CO, combustibles (LEL), H2S, O2, SO2 Multigas Detector; O2/SO2/CO/H2S/LEL, Data Logger, Pump
Call For Price
BW Technologies GasAlertMicro 5 5 Multigas Detector: O2, ClO2, Combustibles, PID WW-10107-68 Mfr # M5PID-XWQV-R-D-0-Y-N-00
COMPARE
Rechargable battery 5 Multigas Detector: O2, ClO2, Combustibles, PID
Call For Price
BW Technologies GasAlertMicro 5 Multigas Detector; O2/ClO2/LEL/PID, Data Logger, Pump WW-10107-69 Mfr # M5PID-XWQV-R-P-D-Y-N-00
COMPARE
Rechargable battery Chlorine dioxide, combustibles (LEL), O2, PID Multigas Detector; O2/ClO2/LEL/PID, Data Logger, Pump
Call For Price
Showing 1 - 20 of 24

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat