This product is now discontinued, View Related Products
Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Observe temperatures of samples when evaporated and condensed under controlled conditions
  • Conforms to ASTM D86, D285, and related ASTM international specifications
Distillation system evaporates and condenses samples under controlled conditions, for observation of the temperature at which various percentages are recovered, and/or the percentages recovered at specified temperatures.
Front View Distillation Apparatus, Groups 0, 1, 2, and 3
Designed for testing motor and aviation gasolines, aviation turbine fuels, napthas, kerosenes, distillate fuels, natural gasoline, liquid hydrocarbon mixtures, and other petroleum products. Features a 1000 W heater with stepless variable control, and push-turn heater elevation control knob.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K45090 Front View Distillation Apparatus, Groups 0, 1, 2, and 3, right side, 220 VAC WW-59873-05 Mfr # K45090
COMPARE
220
Call For Price
Koehler K45100 Front View Distillation Apparatus, Groups 0, 1, 2, and 3, left side, 115 VAC WW-59873-10 Mfr # K45100
COMPARE
115
Call For Price
Koehler K45190 Front View Distillation Apparatus, Groups 0, 1, 2, and 3, left side, 220 VAC WW-59873-15 Mfr # K45190
COMPARE
220