Folding Skirt Stoppers, Rubber or Silicone
Two-part folding skirt stoppers are designed to keep your contents secure two ways. The lower section is molded to grip the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim. Ideal for securing glass, metal, and plastic containers regardless of type. Upper hollow plug section can be perforated to exact contents without removing the stopper — remains watertight even after several perforations.
The rubber stoppers are ideal for chemical transfer and withstand temperatures from -4°F (-20°C) to 212°F (100°C). Chemical-resistent silicone stoppers are recommended for biological applications and withstand temperatures from -76°F (-60°C) to 428°F (220°C)
4,200.00 - ₹16,100.00INR / Pkg of 20
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Top Outside Diameter (OD) (mm)
Availability Pricing
COMPARE
7.5
₹11,400.00INR / Pkg of 100
COMPARE
10.7
₹6,700.00INR / Pkg of 50
COMPARE
13.3
₹8,400.00INR / Pkg of 50
COMPARE
15.5
₹4,200.00INR / Pkg of 20
COMPARE
16.5
₹5,100.00INR / Pkg of 20
COMPARE
17.8
₹6,200.00INR / Pkg of 20
COMPARE
24.4
₹7,300.00INR / Pkg of 10
COMPARE
31.4
₹6,400.00INR / Pkg of 5
COMPARE
₹16,100.00INR / Pkg of 100
COMPARE
10.7
₹9,100.00INR / Pkg of 50
COMPARE
13.3
₹15,700.00INR / Pkg of 50
COMPARE
15.5
₹8,500.00INR / Pkg of 20
COMPARE
16.5
₹8,900.00INR / Pkg of 20
COMPARE
17.8
₹12,800.00INR / Pkg of 20
COMPARE
24.4
₹10,300.00INR / Pkg of 10
COMPARE
31.4
₹8,500.00INR / Pkg of 5
Showing 1- 16 of 16