Fluke® TiS Series Performance Thermal Imagers
Improve performance by seeing the problem and its location in one image with IR-Fusion™ technology
  • One-handed image capture provides ease of use
  • 3.5" LCD screens allow you to get a clear view of your image in the field
  • Smooth auto and manual scaling provide better image quality
  • Durable—IP54-rated housing withstands 2-meter drop
  • Compatible with Fluke Connect™ to instantaneously upload images via Wi-Fi
These innovative thermal imagers feature IR-Fusion™ technology so that you may blend the infrared and visible spectrums together in one image. This helps to identify your problem while simultaneously providing an easy to see location. The imagers can be used one-handed for convenience. You can capture images, review them, and save all with one hand. In addition, Fluke Connect™ offers vast options for analyzing and saving images. Depending on model, the imager can transmit saved images directly to your iPhone®, Android™ 4.3 and up, and direct to PC through a Wi-Fi or LAN connection.
489,000.00 - ₹1,020,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Field Of View (FOV) Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Fluke TiS60+ Performance Series Thermal Imager (320 x 240), 30 HZ, 76,800 Pixels WW-20037-55 Mfr # FLK-TIS60+ 30HZ
COMPARE
1.86 mRad 752 76,800
₹1,020,000.00INR / Each
Fluke TiS20+ Performance Series Thermal Imager (120 x 90), 9 HZ, 10,800 Pixels WW-20037-56 Mfr # FLK-TIS20+ 9HZ
COMPARE
7.6 mRad 302 10,800
₹489,000.00INR / Each