Fluke Oscilloscope Accessories

Upgrade and expand your capabilities
  • Choose among universally compatible and model-specific items
Find test leads, adapters, and much more!
51,500.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Fluke STL120-IV Shielded Test Lead Set, CAT IV 600 V WW-20000-13 Mfr # STL120-IV
COMPARE
Fluke Oscilloscopes Shielded Test Lead Set, CAT IV 600 V
₹51,500.00INR / Each
Fluke VPS41 Voltage Probe Set, 40 MHz, CAT IV 600 V WW-20000-16 Mfr # VPS41
COMPARE
Fluke Oscilloscopes Voltage Probe Set, 40 MHz, CAT IV 600 V
₹51,500.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat