This product is now discontinued, View Related Products
Fluke Calibration 700HPP High-Pressure Pneumatic Test Pump and Pump Kit
Stable enough to use on any surface
  • Generate pneumatic pressures up to 3000 psi (21 MPa)
  • Reaches pressure in 20 seconds to full scale into a 30 cm³ volume
  • Desiccant tube protects against contamination from inlet gas
Use the Fluke Calibration pneumatic test pump to generate pressures up to 3000 psi (20 MPa) without the use of a nitrogen bottle or other external pressure supply. This is an ideal choice for anyone who needs a safe, simple-to-use, portable pressure source they can rely on in a wide variety of conditions. Lightweight pump features collapsible feet, built-in handhold, and comes with a canvas carrying bag making it easily portable. A 2-m (6.5-ft) pressure line and assorted pressure fittings connect a wide variety of DUTs.
Order test pump kit 16105-05 with a calibration manifold to generate high pressure as well as calibrate other devices. The manifold attaches and detaches easily with quick detent pins, so it is easy to set up in the field. A detachable pressure adjustment system and adjustment control knob lets you make fine pressure adjustments to 0.1% of reading or better. An in-line filter system protects the system against contamination from the unit under test. It features a swivel ¼ NPT(F) reference gauge connector that makes switching reference gauges fast and easy.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke Calibration 700HPPK-NPT Pneumatic Test Pump Kit with Manifold WW-16105-05 Mfr # 700HPPK-NPT
COMPARE
Pneumatic Test Pump Kit with Manifold
Call For Price
Fluke Calibration 700HPP-NPT Pneumatic Test Pump WW-16105-06 Mfr # 700HPP-NPT
COMPARE
Pneumatic Test Pump