This product is now discontinued, View Related Products
Flue Gas Analyzers
Unit will not shut off if 15 ppm of carbon monoxide is present for increased safety
  • Pump driven for fast response times
  • Data logging with time stamp
  • Communicate with a PC via the USB output
  • Built-in differential thermometer
  • Attach the combustible gas probe to check valves and fittings for leaks
  • Gas inlet ports allow manometer measurements
  • Store up to 10 sets of readings
  • Stack probe includes a water/filter trap for improved readings
  • Set CO alarm level between 10 and 3000 ppm
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max CO Range (ppm)
Availability Pricing
COMPARE
10000
Call For Price
TPI Flue Gas Analyzer for automotive, industrial, and emission applications WW-10105-29 Mfr # 716N
COMPARE
100000