FLIR® Universal Flexible Current Probes
Simplify current measurements
  • Snake the flexible coil around obstacles — tricky conductors and crowded environments are no longer an issue
  • Connect to your existing clamp meter — the banana plugs are compatible with most DMMs and clamp meters
  • Select your AC current range from 30, 300, or 3000 A
  • See what you''re working on — integrated LEDs illuminate dark cabinets
52,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) AC Min (amps) Description
Availability Pricing
Flir TA74 Universal Flexible Current Probe, 3000 A; 4.7" Conductor Diameter WW-20048-94 Mfr # TA74
COMPARE
3000 30 Universal Flexible Current Probe, 3000 A; 4.7\" Conductor Diameter
₹52,500.00INR / Each