FLIR TG-Series Spot Infrared Thermometers
Measure a smaller targets from a safe distance
  • See the current and last two temperature readings simultaneously on the three-line display
  • Measure smaller areas with laser-tagged precision
  • Rugged, pocket-sized meters can handle up to a 9.8-ft (3-meter) drop
  • Light up dark cabinets with the bright, built-in LED worklight
  • Battery compartment doesn't require tools to open and accepts standard AAA batteries
These spot IR thermometers feature a large, color screen and graphical menu for easy navigation. Plus you can simultaneously view three temperature readings (current and previous two) on the three-line display. Built-in laser indicator and bright LED worklight help you easily identify and see the measurement target—and the distance-to-target ratio allows you to take small area readings, even from a distance.
50,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir TG56 Spot Infrared Thermometer; 30:1 Target Ratio and Type K Input WW-39756-11 Mfr # TG-2
COMPARE
Spot Infrared Thermometer; 30:1 Target Ratio and Type K Input
₹50,000.00INR / Each