This product is now discontinued, View Related Products
FLIR T1K-Series HD Thermal Imaging Cameras
The sharpest results, the truest temperatures, the most flexibility—T1K delivers the quality an expert needs
  • MSX® (multi-spectral dynamic imaging) and UltraMax® for sharper images and thermal clarity
  • Optimized keypad—take advantage of the unique FLIR graphic user interface
  • Highest accuracy for its class—±2%
  • Copy images and reports to USB flash drive
  • Instant, in-camera, multipage report generation
  • Blend thermal and digital images in real time
  • Auto-orientation function
  • Capacitive touch screen
  • T1020-Series cameras add METERLiNK® wireless communication and video with voice annotation
With the highest accuracy for its class at ±2%, and thermal sensitivity as low as 0.045°C, these thermal imagers are perfect for identifying hot/cold spots or a change in temperature, which often indicates a problem. FLIR's thermal imagers are fully radiometric—they measure and store temperatures at every point within an image. Each camera comes with access to robust, web-based, software package allowing the user to transfer images onto a template for analyzing and creating of a report in .pdf format. The T-series also includes built-in software for complete in-field analysis, reporting, and file sharing capabilities—directly from the camera.
FLIR’s UltraMax® and MSX are unique processing techniques that generate reports and images with up to four times as many pixels and 50% less noise than standard native images. More pixel coverage with UltraMax® helps fill in inactive gaps, producing denser temperature measurements for greater thermal accuracy—even from farther away. MSX incorporates real-time thermal video, enhanced with visible spectrum definition, producing exceptional thermal clarity—highlight exactly where the problem is.
Additional features:
• Text and sketch annotation
• Fusion Picture in Picture (PiP)—displays thermal image super-imposed over a digital image
• Thumbnail image gallery—allows quick search of stored images
• Rotating lens—convenient rotating lens detents up to 120° for easy viewing angle
• Li-Ion rechargeable battery—four hours of continuous use
• Laser LocatIR™ pointer—pinpoints the hot spot on the IR image
• Standard SD card—stores more than 1000 radiometric JPEG images
• User-friendly controls—Color touch screen with selectable on-screen viewing modes
• Ruggedly constructed case—IP54 dust/splash-proof
T1020 models add METERLiNK® technology, video recording, and real-time voice annotation. METERLiNK® is a Bluetooth® connection that is an exclusive feature of the FLIR T-Series cameras and select Extech instruments. Measurements from these instruments are transmitted and stored directly on the thermal image and displayed on the camera’s viewing screen. Allowing the user a quick way to review and record multiple parameters at the location being inspected.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Field Of View (FOV) Max Temperature (° F) Description
Availability Pricing
Flir T1020-45 HD Thermal Imaging Camera with METERLiNK®; 45 Degree Lens WW-39756-05 Mfr # FLIR T1020-45
COMPARE
45° × 34° 3632 HD Thermal Imaging Camera with METERLiNK®; 45 Degree Lens
Call For Price
Flir T1020-12 HD Thermal Imaging Camera with METERLiNK®; 12 Degree Lens WW-39756-06 Mfr # FLIR T1020-12
COMPARE
12° × 9° 3632 HD Thermal Imaging Camera with METERLiNK®; 12 Degree Lens
Call For Price
Flir T1010-12 HD Thermal Imaging Camera; 12 Degree Lens WW-39756-88 Mfr # FLIR 1010-12
COMPARE
12° × 9° 1202 HD Thermal Imaging Camera; 12 Degree Lens
Call For Price
Flir T1010-28 HD Thermal Imaging Camera; 28 Degree Lens WW-39756-89 Mfr # FLIR 1010-28
COMPARE
28° × 21° 1202 HD Thermal Imaging Camera; 28 Degree Lens
Call For Price
Flir T1010-45 HD Thermal Imaging Camera; 45 Degree Lens WW-39756-90 Mfr # FLIR 1010-45
COMPARE
45° × 34° 1202 HD Thermal Imaging Camera; 45 Degree Lens
Call For Price
Flir T1020-28 & 12 HD Thermal Imaging Camera with METERLiNK®; 28 and 12 Degree Lens WW-39756-91 Mfr # FLIR 1020-28 & 12
COMPARE
28° × 21°/ 12° × 9° 3632 HD Thermal Imaging Camera with METERLiNK®; 28 and 12 Degree Lens
Call For Price
Flir T1020-28 & 45 HD Thermal Imaging Camera with METERLiNK®; 28 and 45 Degree Lens WW-39756-92 Mfr # FLIR 1020-28 & 45
COMPARE
28° × 21° / 45° × 34° 3632 HD Thermal Imaging Camera with METERLiNK®; 28 and 45 Degree Lens
Call For Price
Flir T1020-28, 12, & 45 HD Thermal Imaging Camera with METERLiNK®; 28, 12, and 45 Degree Lens WW-39756-93 Mfr # FLIR 1020-28 W/ 12 AND 45 LENS
COMPARE
28° × 21° / 45° × 34° / 12° × 9° 3632 HD Thermal Imaging Camera with METERLiNK®; 28, 12, and 45 Degree Lens
Call For Price
Showing 1- 8 of 8