This product is now discontinued, View Related Products
FLIR® Industrial TRMS Multimeters with Type K Temperature
Diagnostic functions provide fast and effective troubleshooting
  • Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
  • Automatically select the optimal measuring range with autoranging feature
  • Bright, large 6000-count 2.8” digital LCD with bargraph
  • CAT IV-600 V, CAT III-1000 V safety rating
These industrial True RMS digital multimeters feature diagnostic measurements including 10 A AC/DC current, 1000 V AC/DC voltage, capacitance, VFD mode, and frequency. Plus, you’ll work more efficiently with advanced features such as diode testing, continuity check, non-contact voltage detection, HOLD function, and a built-in temperature probe for temperature measurement capability. Power your multimeter with alkaline batteries or optional LI-Poly rechargeable battery. For the optional rechargeable battery, order the battery kit separately as part number 20070-01.
These multimeters are CAT III 1000 V and CAT IV 600 V rated and can withstand a drop test up to 3 meters. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically and graphically with analog bar graph on the large, backlit LCD. Min/Max and average readings are recorded to capture variations.
Choose the DM91 model for added Bluetooth® connectivity and datalogging features.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) AC Max (Volts) Description
Availability Pricing
Flir DM90 Industrial TRMS Multimeter with Type K Temperature WW-35725-25 Mfr # DM90
COMPARE
10 1000 Industrial TRMS Multimeter with Type K Temperature
Call For Price
Flir DM91 Industrial TRMS Multimeter with Type K Temperature, Datalogging, and Wireless Connectivity WW-35725-26 Mfr # DM91
COMPARE
10 1000 Industrial TRMS Multimeter with Type K Temperature, Datalogging, and Wireless Connectivity