FLIR® GF77™ Gas Thermal Imaging Camera with Interchangeable Lens
Swappable lens options provide a quick way to visualize gases of interest
  • Gas-specific, spectral filtered lenses can be easily changed out in the field
  • Temperature and gas measurement features can be used simultaneously
  • One tap on the screen automatically adjusts contrast between the gas and the background environment
  • High Sensitivity Mode (HSM) amplifies plume movement for easier gas detection
  • Data reporting is simplified through built-in voice annotation, GPS tagging, customizable work folders, and Wi-Fi connection
This uncooled infrared camera measures temperature and visualizes gas emissions to accurately diagnose electrical and mechanical faults. The ergonomic design has a 180° rotating optical block, touch-screen LCD, and an eyepiece for use in direct sunlight. The camera can be used with multiple lenses that are interchangeable.
The lenses are spectrally filtered from 7 to 8.5 µm and 9.5 to 12 µm, respectively. The low-range (LR) lens are used for detecting methane, nitrous oxide, propane, sulfur dioxide, and refrigerants R-134a or R-152a. The high-range (HR) lens are for detecting sulfur hexafluoride, ammonia, and ethylene. The lenses allow the camera to scan components quickly, focusing on areas with small leaks that may be missed with a traditional TVA (Toxic Vapor Analyzer) or "sniffer". The efficiency of this leak detection protects the environment, lessens product losses, and provides a safer working environment.
9,116,000.00 - ₹15,161,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm Lens WW-39758-16 Mfr # GF77-LR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm Lens
₹9,116,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° High-Range (HR), 9.5-12 µm Lens WW-39758-17 Mfr # GF77-HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° High-Range (HR), 9.5-12 µm Lens
₹9,116,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm Lens WW-39758-18 Mfr # GF77-LR-HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm Lens
₹10,500,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm Lens WW-39758-19 Mfr # GF77-LR-6LR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm Lens
₹11,883,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens WW-39758-20 Mfr # GF77-HR-6HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens
₹11,883,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm, + 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens WW-39758-21 Mfr # GF77-LR-HR-6HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm, + 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens
₹12,410,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm Lens WW-39758-22 Mfr # GF77-LR-6LR-HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm Lens
₹13,266,000.00INR / Each
Flir GF77™ Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens WW-39758-23 Mfr # GF77-LR-6LR-HR-6HR
COMPARE
Gas Thermal Imaging Camera with MSX® and 25° Low-Range (LR), 7-8.5 µm + 6° Low-Range (LR), 7-8.5 µm + 25° High-Range (HR), 9.5-12 µm + 6° High-Range (HR), 9.5-12 µm Lens
₹15,161,000.00INR / Each
Showing 1- 8 of 8