This product is now discontinued, View Related Products
FLIR® Flexible Clamp Meters with Bluetooth® Interface
Get rid of your rigid-jaw clamp meter!
  • Snake the flexible coil around obstacles — tricky conductors and crowded environments are no longer an issue
  • Measure up to 3000 A of True RMS current with these standalone clamp meters
  • Obtain measurements at a safe distance — Bluetooth® capability allows remote viewing
  • See what you''re working on — integrated LEDs illuminate dark cabinets
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Flir CM55 Flexible Clamp Meter, 3000 A; 2.4" Conductor Diameter WW-20048-80 Mfr # CM55
COMPARE
Yes
Call For Price
Flir CM57 Flexible Clamp Meter, 3000 A; 4.7" Conductor Diameter WW-20048-90 Mfr # CM57
COMPARE
Yes