This product is now discontinued, View Related Products
FLIR DM92 Multimeter with VFD Mode
VFD Mode provides additional accuracy and flexibility
  • VFD mode filters noisy variable frequency drives and provides accurate measurements during maintenance and troubleshooting
  • LoZ eliminates ghost voltage errors on long wires sharing conduit
  • Convenient work lights allow you to focus on the test leads without needing a separate flashlight
  • Datalogging allows storage of electrical measurements for future access
  • FLIR Tools APP connects to your compatible mobile device
  • METERLiNK™ technology transmits real-time readings to a METERLiNK™ enabled FLIR infrared thermal imaging camera
This industrial grade digital multimeter includes features for the most demanding measurements on
complex machinery. The rugged design provides durability in challenging environments and helps extend meters overall life. Features include min, max, average recording, auto power off, hold, relative, peak hold, store, recall, and more.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Flir DM92 True RMS Industrial Multimeter with VFD Mode WW-20048-39 Mfr # DM92
COMPARE
1000 ±0.05% Yes