BriskHeat® Flexible Small-Diameter Fiberglass Heating Cords - 240 VAC
Spiral wound heating element-packs more heating element into each inch of cord.
  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
240 VAC models have stripped ends.
Use this table to determine the best coil spacing for you application. For example, to achieve a maximum temperature of 602 ºF (317ºC) on a 1" OD tubing at 120 VAC, you need to space the heating coils 1/4" apart.
Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications. A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord.
Control heating cord temperature with our DIGI-SENSE® temperature controller model 89800-02 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads.
24,600.00 - ₹28,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat CPHTC2012 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 12', 238 Watts, 240 V WW-03122-13 Mfr # CPHTC2012
COMPARE
12 850 240
₹28,200.00INR / Each
BriskHeat CPHTC2030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 240 V WW-03122-31 Mfr # CPHTC2030
COMPARE
3 240
₹25,000.00INR / Each
BriskHeat CPHTC2060 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 6', 136 Watts, 240 V WW-03122-61 Mfr # CPHTC2060
COMPARE
6 240
₹24,600.00INR / Each