BriskHeat® Flexible Small-Diameter Fiberglass Heating Cords - 120 VAC
Spiral wound heating element-packs more heating element into each inch of cord.
  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications.
A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord. Cords feature low power consumption: the 3-,6-, and 12-ft 120 VAC cords (heating 1/4", 1/2", and 1" glass tubing respectively) draw about 0.1 watt/inch at full voltage/high temperature.
Control heating cord temperature with our DIGI-SENSE® temperature controller model 89800-01 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads.
Effective Oct-2014 these heating cords will come with a 120 VAC split plug that came be pluged into a standard oulet. Cords produced before Oct-2014 have a tip plugs and require the power cord adapter 03122-71
24,600.00 - ₹39,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Power (watts) Description
Availability Pricing
BriskHeat CPHTC1012 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 12', 238 Watts, 120 V WW-03122-12 Mfr # CPHTC1012
COMPARE
120 238 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 12', 238 Watts, 120 V
₹28,200.00INR / Each
BriskHeat CPHTC1018 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 18', 416 Watts, 120 V WW-03122-18 Mfr # CPHTC1018
COMPARE
120 416 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 18', 416 Watts, 120 V
₹39,400.00INR / Each
BriskHeat CPHTC1024 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 24', 550 Watts, 120 V WW-03122-24 Mfr # CPHTC1024
COMPARE
120 550 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 24', 550 Watts, 120 V
₹31,300.00INR / Each
BriskHeat CPHTC1030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 120 V WW-03122-30 Mfr # CPHTC1030
COMPARE
120 53 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 120 V
₹25,000.00INR / Each
BriskHeat CPHTC1060 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 6', 136 Watts, 120 V WW-03122-60 Mfr # CPHTC1060
COMPARE
120 136 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 6', 136 Watts, 120 V
₹24,600.00INR / Each