Flanged sanitary gaskets
Hands-free installation for difficult-to-reach locations!
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port -- once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).
230.00 - ₹1,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
Platinum-cured silicone
₹230.00INR / Each
COMPARE
Platinum-cured silicone
₹230.00INR / Each
COMPARE
FKM
₹1,800.00INR / Each
COMPARE
Buna N
₹380.00INR / Each
COMPARE
Buna N
₹370.00INR / Each