Sherwood Scientific Flame Photometer Cleaning and Diluting Solutions

Keep your flame photometer system running smothly
These cleaning and diluting solutions are ideal for keeping your flame photometer in good working order,
16,500.00 - ₹36,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sherwood Scientific Deproteinizing Concentrate for Flame Photometers, 6/pk WW-02655-31 Mfr # 00156182
COMPARE
Deproteinizing Concentrate for Flame Photometers, 6/pk
₹18,000.00INR / Pkg of 6
Sherwood Scientific Cleaning Solution for Flame Photometers, 500 mL WW-02655-79 Mfr # 00156184
COMPARE
Cleaning Solution for Flame Photometers, 500 mL
₹16,500.00INR / Each
Sherwood Scientific 00156682 Tubing Disinfectant for Flame Photometers, 6 x 100 mL WW-02655-81 Mfr # 00156682
COMPARE
Tubing Disinfectant for Flame Photometers, 6 x 100 mL
₹22,500.00INR / Pkg of 1
Sherwood Scientific 00156681 Diluent Concentrate for Flame Photometers, 6 x 100 mL WW-02655-83 Mfr # 00156681
COMPARE
Diluent Concentrate for Flame Photometers, 6 x 100 mL
₹36,000.00INR / Pkg of 6
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat