This product is now discontinued, View Related Products
Fischer Technology Coating Thickness Gauge
Available for sale in the United States, Canada, and Mexico only Portable instruments deliver non-destructive, high precision measurements
  • All models feature:
  • Automatic probe recognition
  • USB port for data transfer to a PC
  • Large display rich in contrast - 240x160 pixels
  • Memory for up to 1000 readings
  • Audible signal at measurement acquisition
  • Displays mean value, standard deviation, min, max, and range
  • Easy adaptation to the shape of the specimen
  • Additional corrective calibration using one or two calibration foils
  • Master calibration for exact characteristics settings for extreme material and geometric properties
  • Capable of storing the master calibration in the connected probe
  • Units of measurement can be switched between µm and mils
  • Adjustable instrument switch-off or continuous operation
  • Low battery indication
  • Lockable keyboard/restricted operating mode
  • Mechanical sliders to cover keys not required for the measurement operation
  • Various language settings
  • NOTE: If purchasing calibration item 17101-69 - Gauges cannot be calibrated without a probe, and probes cannot be calibrated without gauges
  • Probes not included, sold separately
These easy to operate, rugged instruments are suitable for virtually any coating thickness measurement requirement. With a wide assortment of plug-in type probes these meters are designed for non-destructive coating thickness measurements according to the magnetic induction method (Deltascope® FPM10), the eddy current method (Isoscope® FPM10), or both methods in one instrument (Dualscope® FPM20). In addition to one user created calibration application, the spectrum of requirements that this series is capable of spans from extensive painted specimens to other very specific and complex measuring challenges.
226,000.00 - ₹272,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Measuring Method
Availability Pricing
Fischer Technology 605-021 Coating Thickness Gauge WW-54100-00 Mfr # 605-021
COMPARE
Magnetic induction
₹226,000.00INR / Each
Fischer Technology 605-027 Coating Thickness Gauge WW-54100-01 Mfr # 605-027
COMPARE
Eddy current
₹242,000.00INR / Each
Fischer Technology FMP 20 Dualscope Coating Thickness Gauge WW-54100-02 Mfr # 605-023
COMPARE
Eddy current, Magnetic induction
₹272,000.00INR / Each