This product is now discontinued, View Related Products

Filters for 54-Inch Fume Hoods

Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 54” self-monitoring ductless fume hoods
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Exterior Width (in) Filter Type
Availability Pricing
Primary Hydrocarbon Filters for 54" Self-Monitoring Ductless Fume Hoods (2 required) WW-33713-52 Mfr # AU-3804
COMPARE
54 Primary Hydrocarbon
Call For Price
Secondary Hydrocarbon Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-57 Mfr # AU-5704
COMPARE
54 Secondary Hydrocarbon
Call For Price
Primary Mineral Acid Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-62 Mfr # AU-3814
COMPARE
54 Primary Mineral Acid
Call For Price
Secondary Mineral Acid Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-67 Mfr # AU-5714
COMPARE
54 Secondary Mineral Acid
Call For Price
Primary Formaldehyde Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-72 Mfr # AU-3807
COMPARE
54 Primary Formaldehyde
Call For Price
Secondary Formaldehyde Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-77 Mfr # AU-5707
COMPARE
54 Secondary Formaldehyde
Call For Price
Primary HEPA Filters for 54" Self-Monitoring Ductless Fume Hoods (2 required) WW-33713-82 Mfr # AU-3820
COMPARE
54 Primary HEPA
Call For Price
Secondary HEPA Filters for 54" Self-Monitoring Ductless Fume Hoods WW-33713-87 Mfr # AU-5720
COMPARE
54 Secondary HEPA
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat