BriskHeat® Fibrox High-Temperature Heating Tapes leads same ends-120 VAC
Flexible, multipurpose, direct-contact heaters are ideal for use on conductive surfaces
  • Maximum exposure temperature of 900°F (482°C)
  • Choose from power leads on same end or on opposite ends
  • Heavy-duty insulated design
  • Nongrounded
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric.
The 120 VAC tapes include a standard two-prong plug.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
19,100.00 - ₹53,500.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TBIH051-020-LSE Fibrox Insulated Heating Tape Leads On Same End, 2' x 1/2" , 104 Watts, 120 V WW-36110-00 Mfr # TBIH051-020-LSE
COMPARE
2 900 120
₹19,100.00INR / Each
BriskHeat TBIH051-040-LSE Fibrox Insulated Heating Tape Leads On Same End, 4' x 1/2" , 208 Watts, 120 V WW-36110-02 Mfr # TBIH051-040-LSE
COMPARE
4 900 120
₹26,400.00INR / Each
BriskHeat TBIH051-060-LSE Fibrox Insulated Heating Tape Leads On Same End, 6' x 1/2" , 312 Watts, 120 V WW-36110-04 Mfr # TBIH051-060-LSE
COMPARE
6 900 120
₹19,100.00INR / Each
BriskHeat TBIH051-080-LSE Fibrox Insulated Heating Tape Leads On Same End, 8' x 1/2" , 416 Watts, 120 V WW-36110-06 Mfr # TBIH051-080-LSE
COMPARE
8 900 120
₹23,500.00INR / Each
BriskHeat TBIH051-100-LSE Fibrox Insulated Heating Tape Leads On Same End, 10' x 1/2" , 520 Watts, 120 V WW-36110-08 Mfr # TBIH051-100-LSE
COMPARE
10 900 120
₹26,300.00INR / Each
BriskHeat TBIH101-020-LSE Fibrox Insulated Heating Tape Leads On Same End, 2' x 1" , 208 Watts, 120 V WW-36110-10 Mfr # TBIH101-020-LSE
COMPARE
2 900 120
₹22,300.00INR / Each
BriskHeat TBIH101-040-LSE Fibrox Insulated Heating Tape Leads On Same End, 4' x 1" , 416 Watts, 120 V WW-36110-12 Mfr # TBIH101-040-LSE
COMPARE
4 900 120
₹22,300.00INR / Each
BriskHeat TBIH101-060-LSE Fibrox Insulated Heating Tape Leads On Same End, 6' x 1" , 624 Watts, 120 V WW-36110-14 Mfr # TBIH101-060-LSE
COMPARE
6 900 120
₹31,900.00INR / Each
BriskHeat TBIH101-080-LSE Fibrox Insulated Heating Tape Leads On Same End, 8' x 1" , 832 Watts, 120 V WW-36110-16 Mfr # TBIH101-080-LSE
COMPARE
8 900 120
₹34,800.00INR / Each
BriskHeat 1045 Fibrox Insulated Heating Tape Leads On Same End, 120" , Watts, 120V WW-36110-18 Mfr # TBIH-101-100-LSE
COMPARE
10 900 120
₹39,100.00INR / Each
BriskHeat TBIH171-040-LSE Fibrox Insulated Heating Tape Leads On Same End, 4' x 1-3/4" , 416 Watts, 120 V WW-36110-22 Mfr # TBIH171-040-LSE
COMPARE
4 900 120
₹40,800.00INR / Each
BriskHeat TBIH171-060-LSE Fibrox Insulated Heating Tape Leads On Same End, 6' x 1-3/4" , 624 Watts, 120 V WW-36110-24 Mfr # TBIH171-060-LSE
COMPARE
6 900 120
₹47,200.00INR / Each
BriskHeat TBIH171-080-LSE Fibrox Insulated Heating Tape Leads On Same End, 8' x 1-3/4" , 832 Watts, 120 V WW-36110-26 Mfr # TBIH171-080-LSE
COMPARE
8 900 120
₹53,500.00INR / Each
Showing 1- 13 of 13