Fiberglass Trays
Rugged fiberglass trays withstand temperatures 0 to 180°F (-17 to 83°C) under normal usa. Autoclavable to 500°F (204°C) for short intervals.
3,900.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (in) Overall Length (in)
Availability Pricing
COMPARE
3/4 10
₹3,900.00INR / Each